siRNA Details
Gene Symbol: PPP2R2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291310.1 | NP_001278239.1 | 2357 | |||
NM_018461.4 | NP_060931.2 | 1320 | ||||
NR_033191.2 | 1306 | |||||
XM_006717914.3 | XP_006717977.1 | 7, 8 | 1030 | |||
XM_006717915.2 | XP_006717978.1 | 736 | ||||
XM_017016406.1 | XP_016871895.1 | 1382 | ||||
XM_017016407.1 | XP_016871896.1 | 5, 6 | 1382 | |||
XM_017016408.1 | XP_016871897.1 | 588 | ||||
GenBank mRNA | AB040974.1 | - | 1795 | |||
AF220046.1 | - | 3, 4 | 251 | |||
AK000402.1 | - | 4, 5 | 514 | |||
AK293065.1 | - | 7, 8 | 836 | |||
BC047379.1 | - | 7, 8 | 1320 | 5267510 | ||
BC058076.1 | - | 2, 3 | 314 | |||
BC072402.1 | - | 4, 5 | 636 | 6048730 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
