siRNA Details
Gene Symbol: PPP2R2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291310.1 | NP_001278239.1 | 2579 | |||
NM_018461.4 | NP_060931.2 | 1542 | ||||
NR_033191.2 | 1528 | |||||
XM_006717914.3 | XP_006717977.1 | 8 | 1252 | |||
XM_006717915.2 | XP_006717978.1 | 958 | ||||
XM_017016406.1 | XP_016871895.1 | 1604 | ||||
XM_017016407.1 | XP_016871896.1 | 6 | 1604 | |||
XM_017016408.1 | XP_016871897.1 | 810 | ||||
GenBank mRNA | AB040974.1 | - | 2017 | |||
AF220046.1 | - | 4 | 475 | |||
AK000402.1 | - | 5 | 736 | |||
AK001483.1 | - | 1 | 178 | |||
AK293065.1 | - | 8 | 1058 | |||
BC047379.1 | - | 8 | 1542 | 5267510 | ||
BC058076.1 | - | 3 | 536 | |||
BC072402.1 | - | 5 | 858 | 6048730 |

More Information
Other Species Targeted

Gene Ontology Categories:
