siRNA Details
Gene Symbol: SAMD4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303614.1 | NP_001290543.1 | 13 | 2655 | ||
NM_018028.3 | NP_060498.2 | 16 | 3585 | |||
XM_011527063.2 | XP_011525365.1 | 14 | 2874 | |||
XM_011527064.2 | XP_011525366.1 | 14 | 2666 | |||
XM_017026920.1 | XP_016882409.1 | 15 | 2987 | |||
XM_017026921.1 | XP_016882410.1 | 13 | 2850 | |||
XM_017026922.1 | XP_016882411.1 | 14 | 3174 | |||
XM_017026923.1 | XP_016882412.1 | 14 | 8712 | |||
GenBank mRNA | AK091041.1 | - | 2 | 869 | ||
BC039652.1 | - | 9 | 1353 | |||
BC054518.1 | - | 16 | 3576 | 6501489 | ||
BC065211.1 | - | 10 | 1872 | |||
BC080641.1 | - | 13 | 2616 | 6645022 | ||
EF601121.1 | - | 16 | 3572 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
