siRNA Details
Gene Symbol: SAMD4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303614.1 | NP_001290543.1 | 8 | 1828 | ||
NM_018028.3 | NP_060498.2 | 11 | 2549 | |||
XM_011527057.2 | XP_011525359.1 | 9 | 1839 | |||
XM_011527060.2 | XP_011525362.1 | 9 | 2941 | |||
XM_011527061.2 | XP_011525363.1 | 8 | 1753 | |||
XM_011527062.2 | XP_011525364.1 | 8 | 1814 | |||
XM_011527063.2 | XP_011525365.1 | 9 | 1838 | |||
XM_011527064.2 | XP_011525366.1 | 9 | 1839 | |||
XM_017026918.1 | XP_016882407.1 | 8 | 1773 | |||
XM_017026919.1 | XP_016882408.1 | 10 | 1929 | |||
XM_017026920.1 | XP_016882409.1 | 10 | 1951 | |||
XM_017026921.1 | XP_016882410.1 | 8 | 1814 | |||
XM_017026922.1 | XP_016882411.1 | 10 | 1931 | |||
XM_017026923.1 | XP_016882412.1 | 10 | 7469 | |||
GenBank mRNA | BC039652.1 | - | 4 | 526 | ||
BC054518.1 | - | 11 | 2540 | 6501489 | ||
BC065211.1 | - | 5 | 836 | |||
BC080641.1 | - | 8 | 1788 | 6645022 | ||
EF601121.1 | - | 11 | 2536 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
