siRNA Details
Gene Symbol: RBMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002897.4 | NP_002888.1 | 14 | 3159 | ||
NM_016836.3 | NP_058520.1 | 14 | 3168 | |||
XM_005246737.1 | XP_005246794.1 | 15 | 2927 | |||
XM_005246738.2 | XP_005246795.1 | 15 | 3262 | |||
XM_005246739.2 | XP_005246796.1 | 15 | 3132 | |||
XM_005246740.1 | XP_005246797.1 | 15 | 2918 | |||
XM_005246741.3 | XP_005246798.1 | 15 | 3129 | |||
XM_017004626.1 | XP_016860115.1 | 14 | 3205 | |||
GenBank mRNA | AL117617.1 | - | 1 | 210 | ||
BC065192.1 | - | 1 | 429 | |||
BC069687.1 | - | 1 | 376 | |||
BC069706.1 | - | 1 | 375 | |||
BC069763.1 | - | 1 | 375 | |||
BC080620.1 | - | 1 | 404 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
