siRNA Details
Gene Symbol: TPPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015964.3 | NP_057048.2 | 4 | 1019 | ||
NM_016140.3 | NP_057224.2 | 5 | 1110 | |||
GenBank mRNA | AF078846.1 | - | 4 | 950 | ||
AF132972.1 | - | 4 | 938 | |||
BC000691.2 | - | 4 | 966 | 3349849 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
