siRNA Details
Gene Symbol: COPG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016128.3 | NP_057212.1 | 21 | 2300 | ||
GenBank mRNA | AB047846.1 | - | 21 | 2215 | ||
AF100756.1 | - | 21 | 2275 | |||
AK001724.1 | - | 21 | 2278 | |||
AK024721.1 | - | 8 | 774 | |||
AK025147.1 | - | 10 | 1256 | |||
AK123939.1 | - | 17 | 1867 | |||
AK291693.1 | - | 21 | 2304 | |||
AK309821.1 | - | 20 | 2459 | |||
BC020498.1 | - | 14 | 1676 | |||
BC066650.1 | - | 21 | 2263 | 6046736 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
