siRNA Details
Gene Symbol: COPG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016128.3 | NP_057212.1 | 10 | 897 | ||
XM_011512549.2 | XP_011510851.1 | 10 | 932 | |||
GenBank mRNA | AB047846.1 | - | 10 | 812 | ||
AF100756.1 | - | 10 | 872 | |||
AK001724.1 | - | 10 | 875 | |||
AK123939.1 | - | 6 | 464 | |||
AK291693.1 | - | 10 | 901 | |||
AK309821.1 | - | 9 | 1056 | |||
BC020498.1 | - | 3 | 273 | |||
BC066650.1 | - | 10 | 860 | 6046736 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
