siRNA Details
Gene Symbol: C5orf51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175921.5 | NP_787117.3 | 6 | 4042 | ||
XM_005248289.4 | XP_005248346.1 | 5 | 3958 | |||
XM_011514032.2 | XP_011512334.1 | 7 | 4171 | |||
GenBank mRNA | AK094002.1 | - | 1 | 878 | ||
AL833916.1 | - | 1 | 1144 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
