siRNA Details
Gene Symbol: PVT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003367.3 | 1 | 180 | |||
GenBank mRNA | BM821521.1 | - | 1 | 180 | ||
BX280136.1 | - | 1 | 159 | |||
M25802.1 | - | 1 | 154 | |||
M25803.1 | - | 1 | 151 | |||
M34428.1 | - | 1 | 154 | |||
M34431.1 | - | 1 | 155 |
Gene Symbol: LINC00824 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC014119.1 | - | 163 | |||
BQ223316.1 | - | 166 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
