siRNA Details
Gene Symbol: KRTAP3-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033185.2 | NP_149441.1 | 1 | 620 | ||
GenBank mRNA | AJ406933.1 | - | 1 | 620 | ||
BC069099.1 | - | 1 | 620 | 7216913 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
