siRNA Details
Gene Symbol: CDC26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139286.3 | NP_644815.1 | 4 | 672 | ||
XM_017014573.1 | XP_016870062.1 | 4 | 639 | |||
XM_017014574.1 | XP_016870063.1 | 4 | 1104 | |||
XM_017014575.1 | XP_016870064.1 | 4 | 634 | |||
GenBank mRNA | BC042534.1 | - | 4 | 627 | 4751173 | |
BC066300.1 | - | 4 | 657 | 5258904 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
