siRNA Details
Gene Symbol: MED29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317770.1 | NP_001304699.1 | 4 | 2141 | ||
NM_017592.2 | NP_060062.1 | 4 | 2211 | |||
NR_133915.1 | 3 | 2152 | ||||
GenBank mRNA | AK000411.1 | - | 4 | 2121 | ||
AK026527.1 | - | 1 | 630 | |||
AK090877.1 | - | 3 | 2043 | |||
AK092909.1 | - | 3 | 1964 | |||
AK124629.1 | - | 5 | 2074 | |||
AL137304.1 | - | 4 | 2126 | |||
AY729650.1 | - | 4 | 2191 | |||
BC019015.2 | - | 4 | 2118 | 4129619 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
