siRNA Details
Gene Symbol: GPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004489.4 | NP_004480.1 | 6 | 601 | ||
GenBank mRNA | AK301877.1 | - | 7 | 613 | ||
BC013652.1 | - | 6 | 601 | |||
BC103901.1 | - | 7 | 826 | 40005162 | ||
BC103902.1 | - | 5 | 1371 | |||
BC103903.1 | - | 7 | 826 | 40005170 | ||
BC107738.1 | - | 7 | 636 | 5810755 | ||
BT006998.1 | - | 6 | 525 | |||
CR541723.1 | - | 6 | 525 | |||
CR541750.1 | - | 6 | 525 | |||
U28963.1 | - | 7 | 615 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
