siRNA Details
Gene Symbol: GPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004489.4 | NP_004480.1 | 2 | 195 | ||
GenBank mRNA | AK301877.1 | - | 3 | 207 | ||
BC013652.1 | - | 2 | 195 | |||
BC103901.1 | - | 3 | 420 | 40005162 | ||
BC103902.1 | - | 2 | 781 | |||
BC103903.1 | - | 3 | 420 | 40005170 | ||
BC107738.1 | - | 3 | 230 | 5810755 | ||
BT006998.1 | - | 2 | 119 | |||
CR541723.1 | - | 2 | 119 | |||
CR541750.1 | - | 2 | 119 | |||
U28963.1 | - | 3 | 209 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
