siRNA Details
Gene Symbol: NEURL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005408.1 | NP_001005408.1 | 11 | 2022 | ||
NM_032442.2 | NP_115818.2 | 11 | 2022 | |||
GenBank mRNA | AB058690.1 | - | 11 | 2022 | ||
AL136870.1 | - | 11 | 1878 | |||
AL833570.1 | - | 11 | 1268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
