siRNA Details
Gene Symbol: NEURL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032442.2 | NP_115818.2 | 15, 16 | 2606 | ||
GenBank mRNA | AB058690.1 | - | 15, 16 | 2606 | ||
AL833570.1 | - | 15, 16 | 1852 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
