siRNA Details
Gene Symbol: MROH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288814.1 | NP_001275743.1 | 42 | 4953 | ||
NM_032450.2 | NP_115826.2 | 44 | 5093 | |||
XM_011517270.2 | XP_011515572.1 | 45 | 5285 | |||
XM_011517271.2 | XP_011515573.1 | 43 | 5297 | |||
XM_011517272.2 | XP_011515574.1 | 44 | 5258 | |||
XM_011517273.2 | XP_011515575.1 | 44 | 5229 | |||
XM_011517274.2 | XP_011515576.1 | 43 | 5202 | |||
XM_011517275.2 | XP_011515577.1 | 43 | 5040 | |||
XM_011517276.2 | XP_011515578.1 | 45 | 5311 | |||
XM_011517277.1 | XP_011515579.1 | 45 | 5147 | |||
XM_011517278.1 | XP_011515580.1 | 44 | 5027 | |||
XM_011517279.2 | XP_011515581.1 | 44 | 5191 | |||
XM_011517281.1 | XP_011515583.1 | 44 | 5200 | |||
XM_011517282.1 | XP_011515584.1 | 43 | 5125 | |||
XM_011517283.2 | XP_011515585.1 | 45 | 5106 | |||
XM_017013802.1 | XP_016869291.1 | 42 | 5269 | |||
XM_017013803.1 | XP_016869292.1 | 42 | 5012 | |||
XM_017013804.1 | XP_016869293.1 | 44 | 5120 | |||
XM_017013805.1 | XP_016869294.1 | 43 | 5000 | |||
XM_017013806.1 | XP_016869295.1 | 42 | 4762 | |||
GenBank mRNA | AK092861.1 | - | 14 | 2189 | ||
AL834145.1 | - | 26 | 3418 | |||
AL834309.1 | - | 43 | 4977 | |||
BC017118.1 | - | 12 | 1530 | |||
CN333563.1 | - | 5 | 447 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
