siRNA Details
Gene Symbol: MROH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288814.1 | NP_001275743.1 | 41 | 4896 | ||
NM_032450.2 | NP_115826.2 | 43 | 5036 | |||
XM_011517270.2 | XP_011515572.1 | 44 | 5228 | |||
XM_011517271.2 | XP_011515573.1 | 42 | 5240 | |||
XM_011517272.2 | XP_011515574.1 | 43 | 5201 | |||
XM_011517273.2 | XP_011515575.1 | 43 | 5172 | |||
XM_011517274.2 | XP_011515576.1 | 42 | 5145 | |||
XM_011517275.2 | XP_011515577.1 | 42 | 4983 | |||
XM_011517276.2 | XP_011515578.1 | 44 | 5254 | |||
XM_011517277.1 | XP_011515579.1 | 44 | 5090 | |||
XM_011517278.1 | XP_011515580.1 | 43 | 4970 | |||
XM_011517279.2 | XP_011515581.1 | 43 | 5134 | |||
XM_011517281.1 | XP_011515583.1 | 43 | 5143 | |||
XM_011517282.1 | XP_011515584.1 | 42 | 5068 | |||
XM_011517283.2 | XP_011515585.1 | 44 | 5049 | |||
XM_017013802.1 | XP_016869291.1 | 41 | 5212 | |||
XM_017013803.1 | XP_016869292.1 | 41 | 4955 | |||
XM_017013804.1 | XP_016869293.1 | 43 | 5063 | |||
XM_017013805.1 | XP_016869294.1 | 42 | 4943 | |||
XM_017013806.1 | XP_016869295.1 | 41 | 4705 | |||
GenBank mRNA | AK092861.1 | - | 13 | 2132 | ||
AL834145.1 | - | 25 | 3361 | |||
AL834309.1 | - | 42 | 4920 | |||
BC017118.1 | - | 11 | 1453 | |||
CN333563.1 | - | 4 | 390 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
