siRNA Details
Gene Symbol: SLC41A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008485.1 | NP_001008485.1 | 8 | 1172 | ||
NM_001008486.1 | NP_001008486.1 | 7 | 1064 | |||
NM_001008487.1 | NP_001008487.1 | 7 | 1091 | |||
NM_001164475.1 | NP_001157947.1 | 7 | 793 | |||
NM_017836.3 | NP_060306.3 | 8 | 1172 | |||
XM_005247559.1 | XP_005247616.1 | 8 | 1050 | |||
XM_005247562.1 | XP_005247619.1 | 8 | 1066 | |||
XM_005247563.2 | XP_005247620.1 | 8 | 1095 | |||
XM_005247564.1 | XP_005247621.1 | 8 | 1050 | |||
XM_005247565.2 | XP_005247622.1 | 7 | 1154 | |||
XM_006713681.2 | XP_006713744.1 | 8 | 1012 | |||
XM_011512945.1 | XP_011511247.1 | 7 | 1199 | |||
XM_017006705.1 | XP_016862194.1 | 8 | 1039 | |||
XM_017006706.1 | XP_016862195.1 | 8 | 1066 | |||
XM_017006707.1 | XP_016862196.1 | 8 | 1089 | |||
XM_017006708.1 | XP_016862197.1 | 7 | 897 | |||
XM_017006709.1 | XP_016862198.1 | 7 | 952 | |||
GenBank mRNA | AK000480.1 | - | 7 | 895 | ||
AK021925.1 | - | 8 | 858 | |||
AK022780.1 | - | 7 | 893 | |||
AK093380.1 | - | 7 | 1088 | |||
AK094051.1 | - | 3 | 1181 | |||
AK128054.1 | - | 3 | 3206 | |||
AK298070.1 | - | 7 | 802 | |||
AW938624.1 | - | 1 | 48 | |||
BC009039.2 | - | 8 | 1005 | 3852588 | ||
BC009444.1 | - | 8 | 990 | |||
BC028241.1 | - | 7 | 988 | 5243239 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
