siRNA Details
Gene Symbol: SH3PXD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014631.2 | NP_055446.2 | 13 | 1438 | ||
XM_005270294.4 | XP_005270351.1 | 14 | 1571 | |||
XM_005270295.4 | XP_005270352.1 | 13 | 1525 | |||
XM_005270297.3 | XP_005270354.1 | 8 | 2802 | |||
XM_005270298.4 | XP_005270355.1 | 8 | 1605 | |||
XM_006718079.3 | XP_006718142.1 | 15 | 1668 | |||
XM_006718080.3 | XP_006718143.1 | 9 | 1699 | |||
XM_011540385.2 | XP_011538687.1 | 14 | 1622 | |||
XM_011540386.2 | XP_011538688.1 | 9 | 2898 | |||
XM_011540387.2 | XP_011538689.1 | 5 | 4544 | |||
XM_017016969.1 | XP_016872458.1 | 14 | 1589 | |||
XM_017016970.1 | XP_016872459.1 | 13 | 1405 | |||
GenBank mRNA | AB007878.1 | - | 11 | 1188 | ||
AK056469.1 | - | 12 | 1161 | |||
AK294454.1 | - | 8 | 978 | |||
AK294736.1 | - | 8 | 1016 | |||
AK295660.1 | - | 8 | 1082 | |||
AK302015.1 | - | 11 | 1194 | |||
AK304828.1 | - | 8 | 994 | |||
AK315983.1 | - | 8 | 1021 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
