siRNA Details
Gene Symbol: SNX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013321.3 | NP_037453.1 | 11 | 1613 | ||
XM_011515329.2 | XP_011513631.1 | 11 | 1726 | |||
XM_011515330.1 | XP_011513632.1 | 11 | 1615 | |||
XM_017012083.1 | XP_016867572.1 | 11 | 1683 | |||
XM_017012084.1 | XP_016867573.1 | 11 | 1725 | |||
GenBank mRNA | AL713633.1 | - | 1 | 92 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
