siRNA Details
Gene Symbol: SPATA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125327.1 | 9 | 1462 | |||
NR_125328.1 | 8 | 1135 | ||||
NR_125330.1 | 8 | 1234 | ||||
GenBank mRNA | AK093225.1 | - | 8 | 1234 | ||
AK292338.1 | - | 9 | 1443 | |||
AK310260.1 | - | 8 | 1116 | |||
AY027526.1 | - | 9 | 1411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
