siRNA Details
Gene Symbol: AKAP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016248.3 | NP_057332.1 | 8 | 3464 | ||
XM_005266247.1 | XP_005266304.1 | 8 | 3465 | |||
XM_005266248.3 | XP_005266305.1 | 7 | 3358 | |||
XM_005266249.3 | XP_005266306.1 | 8 | 3465 | |||
XM_005266250.3 | XP_005266307.1 | 8 | 3465 | |||
XM_011534903.2 | XP_011533205.1 | 8 | 3461 | |||
XM_011534904.1 | XP_011533206.1 | 8 | 3657 | |||
XM_011534905.1 | XP_011533207.1 | 7 | 3415 | |||
XM_011534906.2 | XP_011533208.1 | 7 | 3633 | |||
XM_017020381.1 | XP_016875870.1 | 8 | 3515 | |||
XM_017020382.1 | XP_016875871.1 | 6 | 3305 | |||
XM_017020383.1 | XP_016875872.1 | 6 | 3493 | |||
GenBank mRNA | AB014529.2 | - | 1 | 1404 | ||
AF176555.1 | - | 8 | 3464 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
