siRNA Details
Gene Symbol: PAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018440.3 | NP_060910.3 | 9 | 7946 | ||
XM_017013640.1 | XP_016869129.1 | 8 | 7899 | |||
XM_017013641.1 | XP_016869130.1 | 8 | 7498 | |||
XM_017013642.1 | XP_016869131.1 | 8 | 7511 | |||
XM_017013643.1 | XP_016869132.1 | 8 | 7487 | |||
GenBank mRNA | AK095177.1 | - | 1 | 313 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
