siRNA Details
Gene Symbol: KLHL29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052920.1 | NP_443152.1 | 5 | 1187 | ||
XM_006711929.3 | XP_006711992.1 | 4 | 2283 | |||
XM_017003264.1 | XP_016858753.1 | 4 | 1087 | |||
GenBank mRNA | AK024168.1 | - | 1 | 298 | ||
AL834515.1 | - | 3 | 438 | |||
BC013982.1 | - | 2 | 176 | |||
BC015667.2 | - | 1 | 199 | |||
BC069271.1 | - | 4 | 761 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
