siRNA Details
Gene Symbol: UNCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080461.1 | NP_001073930.1 | 2 | 382 | ||
GenBank mRNA | KM587717.1 | - | 2 | 1018 | ||
KM587718.1 | - | 2 | 552 | |||
KM587719.1 | - | 2 | 552 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
