siRNA Details
Gene Symbol: PRR26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027151.1 | 2 | 271 | |||
NR_027152.2 | 1 | 254 | ||||
GenBank mRNA | AK096000.1 | - | 2 | 271 | ||
AK096013.1 | - | 1 | 254 | |||
AK296168.1 | - | 2 | 186 | |||
BC042561.1 | - | 1 | 65 | |||
BC144473.1 | - | 1 | 354 | |||
DA621677.1 | - | 2 | 271 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
