siRNA Details
Gene Symbol: ABHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032604.3 | NP_115993.3 | 5 | 718 | ||
XM_011533135.2 | XP_011531437.1 | 5 | 684 | |||
XM_011533136.2 | XP_011531438.1 | 5 | 684 | |||
XM_011533137.2 | XP_011531439.1 | 5 | 684 | |||
XM_011533138.2 | XP_011531440.1 | 4 | 566 | |||
XM_017005113.1 | XP_016860602.1 | 4 | 638 | |||
GenBank mRNA | AK093447.1 | - | 5 | 684 | ||
AY033290.1 | - | 5 | 591 | |||
BC039576.1 | - | 5 | 672 | 5744853 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
