siRNA Details
Gene Symbol: ABHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032604.3 | NP_115993.3 | 1 | 252 | ||
XM_011533135.2 | XP_011531437.1 | 1 | 218 | |||
XM_011533136.2 | XP_011531438.1 | 1 | 218 | |||
XM_011533137.2 | XP_011531439.1 | 1 | 218 | |||
XM_011533138.2 | XP_011531440.1 | 1 | 261 | |||
XM_017005113.1 | XP_016860602.1 | 1 | 218 | |||
GenBank mRNA | AK074615.1 | - | 1 | 99 | ||
AK093447.1 | - | 1 | 218 | |||
AY033290.1 | - | 1 | 125 | |||
BC039576.1 | - | 1 | 206 | 5744853 | ||
DB455979.1 | - | 1 | 283 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_021304, AA467032, CK791402, CK789386, AK162873, and others.
Gene Ontology Categories:
