siRNA Details
Gene Symbol: ZNF267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265588.1 | NP_001252517.1 | 4 | 514 | ||
NM_003414.5 | NP_003405.3 | 3 | 363 | |||
NR_049749.1 | 3 | 363 | ||||
XM_017022865.1 | XP_016878354.1 | 4 | 450 | |||
XM_017022866.1 | XP_016878355.1 | 4 | 390 | |||
GenBank mRNA | AF220492.1 | - | 3 | 316 | ||
AK292573.1 | - | 3 | 188 | |||
BC036367.1 | - | 3 | 287 | 4828269 | ||
BC127088.1 | - | 3 | 286 | 40123276 | ||
BC127089.1 | - | 3 | 286 | 40123277 | ||
DA657367.1 | - | 3 | 314 | |||
DB091947.1 | - | 4 | 440 | |||
X78925.1 | - | 2 | 121 |
Gene Symbol: ZNF720 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130913.1 | NP_001124385.1 | 3 | 358 | ||
GenBank mRNA | AK299345.1 | - | 3 | 318 | ||
AL832197.1 | - | 3 | 355 | |||
BC018134.1 | - | 3 | 313 | 3912539 | ||
BC055408.1 | - | 3 | 353 | 3684126 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
