siRNA Details
Gene Symbol: GPM6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001994.2 | NP_001001994.1 | 3 | 541 | ||
NM_001001995.2 | NP_001001995.1 | 4 | 859 | |||
NM_001001996.2 | NP_001001996.1 | 4 | 859 | |||
NM_001318729.1 | NP_001305658.1 | 3 | 541 | |||
NM_005278.4 | NP_005269.1 | 3 | 739 | |||
XM_005274489.4 | XP_005274546.1 | 3 | 738 | |||
XM_011545497.2 | XP_011543799.1 | 4 | 550 | |||
XM_017029432.1 | XP_016884921.1 | 4 | 549 | |||
XM_017029433.1 | XP_016884922.1 | 3 | 243 | |||
XM_017029434.1 | XP_016884923.1 | 3 | 266 | |||
GenBank mRNA | AB209525.1 | - | 3 | 260 | ||
AK095657.1 | - | 4 | 613 | |||
AK294314.1 | - | 3 | 312 | |||
AK299675.1 | - | 5 | 607 | |||
BC008151.1 | - | 3 | 365 | 4215416 | ||
BC047295.1 | - | 4 | 569 | 4215095 | ||
BI599457.1 | - | 4 | 645 | |||
CD516484.1 | - | 3 | 541 | |||
U45955.1 | - | 3 | 339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
