siRNA Details
Gene Symbol: GPM6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001994.2 | NP_001001994.1 | 2, 3 | 516 | ||
NM_001001995.2 | NP_001001995.1 | 3, 4 | 834 | |||
NM_001001996.2 | NP_001001996.1 | 3, 4 | 834 | |||
NM_001318729.1 | NP_001305658.1 | 2, 3 | 516 | |||
NM_005278.4 | NP_005269.1 | 2, 3 | 714 | |||
XM_005274489.4 | XP_005274546.1 | 2, 3 | 713 | |||
XM_011545497.2 | XP_011543799.1 | 3, 4 | 525 | |||
XM_017029432.1 | XP_016884921.1 | 3, 4 | 524 | |||
XM_017029433.1 | XP_016884922.1 | 2, 3 | 218 | |||
XM_017029434.1 | XP_016884923.1 | 2, 3 | 241 | |||
GenBank mRNA | AB209525.1 | - | 2, 3 | 235 | ||
AK095657.1 | - | 3, 4 | 588 | |||
AK294314.1 | - | 2, 3 | 287 | |||
AK299675.1 | - | 4, 5 | 582 | |||
BC008151.1 | - | 2, 3 | 340 | 4215416 | ||
BC047295.1 | - | 3, 4 | 544 | 4215095 | ||
BF525674.1 | - | 2, 3 | 429 | |||
BI599457.1 | - | 3, 4 | 620 | |||
CD516484.1 | - | 2, 3 | 516 | |||
U45955.1 | - | 2, 3 | 314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
