siRNA Details
Gene Symbol: PYCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013328.3 | NP_037460.2 | 4 | 611 | ||
GenBank mRNA | AF087859.1 | - | 4 | 529 | ||
AF151351.1 | - | 4 | 408 | |||
AK001508.1 | - | 2 | 1990 | |||
AK291913.1 | - | 4 | 446 | |||
AL834217.1 | - | 2 | 608 | |||
BC014868.1 | - | 4 | 502 | 3919161 | ||
BC020553.1 | - | 4 | 536 | 2901360 | ||
KU178615.1 | - | 4 | 381 | |||
KU178616.1 | - | 4 | 205 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
