siRNA Details
Gene Symbol: OR52E4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005165.1 | NP_001005165.1 | 1 | 585 | ||
GenBank mRNA | BC136972.1 | - | 1 | 607 | 9020964 | |
BC136974.1 | - | 1 | 607 | 9020966 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
