siRNA Details
Gene Symbol: TBX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000192.3 | NP_000183.2 | 9 | 1803 | ||
NM_080717.2 | NP_542448.1 | 8 | 1618 | |||
NM_181486.2 | NP_852259.1 | 9 | 1710 | |||
XM_017019912.1 | XP_016875401.1 | 9 | 3554 | |||
GenBank mRNA | AB051068.1 | - | 8 | 1562 | ||
AF221714.1 | - | 9 | 1801 | |||
AK290611.1 | - | 9 | 1307 | |||
AK300251.1 | - | 7 | 1104 | |||
BC027942.1 | - | 9 | 1631 | 5204163 | ||
U89353.1 | - | 8 | 1136 | |||
Y09445.1 | - | 9 | 1792 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
