siRNA Details
Gene Symbol: TBX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000192.3 | NP_000183.2 | 3 | 839 | ||
NM_080717.2 | NP_542448.1 | 2 | 654 | |||
NM_181486.2 | NP_852259.1 | 3 | 746 | |||
XM_017019912.1 | XP_016875401.1 | 3 | 2590 | |||
GenBank mRNA | AB051068.1 | - | 2 | 599 | ||
AF221714.1 | - | 3 | 837 | |||
AK290611.1 | - | 3 | 343 | |||
BC027942.1 | - | 3 | 667 | 5204163 | ||
DA863182.1 | - | 3 | 443 | |||
U80987.1 | - | 2 | 172 | |||
U89353.1 | - | 2 | 172 | |||
Y09445.1 | - | 3 | 831 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
