siRNA Details
Gene Symbol: FOXN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085471.1 | NP_001078940.1 | 7 | 6619 | ||
NM_005197.3 | NP_005188.2 | 6 | 6670 | |||
GenBank mRNA | AF116643.2 | - | 1 | 544 | ||
AK123929.1 | - | 1 | 1295 | |||
AK314098.1 | - | 1 | 605 | |||
BC010460.1 | - | 1 | 410 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
