siRNA Details
Gene Symbol: DEFB124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037500.1 | NP_001032589.1 | 2 | 182 | ||
XM_017027782.1 | XP_016883271.1 | 3 | 799 | |||
GenBank mRNA | AY122476.1 | - | 1 | 117 | ||
DQ119827.1 | - | 2 | 182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
