siRNA Details
Gene Symbol: OR2M7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004691.1 | NP_001004691.1 | 1 | 132 | ||
GenBank mRNA | BC136944.1 | - | 1 | 161 | 9020936 | |
BC136946.1 | - | 1 | 161 | 9020938 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
