siRNA Details
Gene Symbol: OR2M2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004688.1 | NP_001004688.1 | 1 | 132 |
Gene Symbol: OR2M3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004689.1 | NP_001004689.1 | 1 | 132 | ||
GenBank mRNA | BC136970.1 | - | 1 | 170 | 9020962 | |
BC136971.1 | - | 1 | 170 | 9020963 |
Gene Symbol: OR2M5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004690.1 | NP_001004690.1 | 1 | 132 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
