siRNA Details
Gene Symbol: LOC646383 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK091840.1 | - | 5 | 575 | ||
BC144418.1 | - | 2 | 159 | |||
BC144419.1 | - | 2 | 159 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
