siRNA Details
Gene Symbol: CBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005188.3 | NP_005179.2 | 16 | 10142 | ||
XM_017018511.1 | XP_016874000.1 | 15 | 9577 | |||
GenBank mRNA | AL832396.1 | - | 1 | 1350 | ||
BX648139.1 | - | 1 | 4894 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
