siRNA Details
Gene Symbol: KRTAP9-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190460.1 | NP_001177389.1 | 1 | 356 | ||
XM_005257633.1 | XP_005257690.1 | 1 | 356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
