siRNA Details
Gene Symbol: MSLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177355.2 | NP_001170826.1 | 15 | 1576 | ||
NM_005823.5 | NP_005814.2 | 15 | 1636 | |||
NM_013404.4 | NP_037536.2 | 14 | 1603 | |||
XM_005255034.4 | XP_005255091.1 | 14 | 1882 | |||
XM_011522348.2 | XP_011520650.1 | 15 | 1673 | |||
XM_017022853.1 | XP_016878342.1 | 14 | 1820 | |||
XM_017022854.1 | XP_016878343.1 | 14 | 1831 | |||
XM_017022855.1 | XP_016878344.1 | 14 | 1729 | |||
XM_017022856.1 | XP_016878345.1 | 14 | 1730 | |||
XM_017022857.1 | XP_016878346.1 | 16 | 1750 | |||
XM_017022858.1 | XP_016878347.1 | 15 | 1670 | |||
GenBank mRNA | AF180951.1 | - | 4 | 580 | ||
AY743922.1 | - | 15 | 1636 | |||
BC003512.1 | - | 14 | 1882 | 3611296 | ||
BC009272.2 | - | 15 | 1575 | 3957372 | ||
D49441.1 | - | 14 | 1560 | |||
U40434.1 | - | 14 | 1585 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
