siRNA Details
Gene Symbol: MSLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177355.2 | NP_001170826.1 | 5 | 277 | ||
NM_005823.5 | NP_005814.2 | 5 | 337 | |||
NM_013404.4 | NP_037536.2 | 4 | 280 | |||
XM_005255034.4 | XP_005255091.1 | 4 | 583 | |||
XM_011522348.2 | XP_011520650.1 | 5 | 350 | |||
XM_017022853.1 | XP_016878342.1 | 4 | 497 | |||
XM_017022854.1 | XP_016878343.1 | 4 | 508 | |||
XM_017022855.1 | XP_016878344.1 | 4 | 430 | |||
XM_017022856.1 | XP_016878345.1 | 4 | 431 | |||
XM_017022857.1 | XP_016878346.1 | 6 | 427 | |||
XM_017022858.1 | XP_016878347.1 | 5 | 347 | |||
GenBank mRNA | AL545401.3 | - | 4 | 555 | ||
AY743922.1 | - | 5 | 337 | |||
BC003512.1 | - | 4 | 583 | 3611296 | ||
BC009272.2 | - | 5 | 276 | 3957372 | ||
D49441.1 | - | 4 | 261 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
