siRNA Details
Gene Symbol: LRRC75B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207644.2 | NP_997527.2 | 3 | 442 | ||
GenBank mRNA | AK097054.1 | - | 3 | 2158 | ||
BC017595.1 | - | 2 | 690 | |||
BC032388.1 | - | 3 | 403 | |||
BC108684.1 | - | 3 | 444 | 6471196 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
