siRNA Details
Gene Symbol: LRRC75B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207644.2 | NP_997527.2 | 2, 3 | 339 | ||
GenBank mRNA | BC032388.1 | - | 2, 3 | 300 | ||
BC108684.1 | - | 2, 3 | 341 | 6471196 | ||
BI544432.1 | - | 2, 3 | 351 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
