siRNA Details
Gene Symbol: OR13C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004481.1 | NP_001004481.1 | 1 | 460 | ||
GenBank mRNA | BC136824.1 | - | 1 | 503 | 9020815 | |
BC136826.1 | - | 1 | 503 | 9020817 |
Gene Symbol: OR13C5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004482.1 | NP_001004482.1 | 1 | 460 | ||
GenBank mRNA | BC136842.1 | - | 1 | 554 | 9020833 | |
BC136843.1 | - | 1 | 554 | 9020834 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
