siRNA Details
Gene Symbol: RNF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201370.1 | NP_001188299.1 | 2 | 374 | ||
NM_014245.4 | NP_055060.1 | 3 | 422 | |||
NM_183237.2 | NP_899060.1 | 3 | 415 | |||
NR_037702.1 | 4 | 686 | ||||
NR_037703.1 | 4 | 679 | ||||
GenBank mRNA | AF092878.1 | - | 3 | 289 | ||
AF142060.1 | - | 3 | 289 | |||
AF164679.1 | - | 3 | 342 | |||
AF312226.1 | - | 4 | 553 | |||
AK311982.1 | - | 3 | 354 | |||
BC005966.1 | - | 3 | 315 | 4069078 | ||
BC008627.1 | - | 3 | 325 | 4177613 | ||
BG715867.1 | - | 3 | 361 | |||
BG716023.1 | - | 3 | 361 | |||
BG720797.1 | - | 4 | 574 | |||
BG742338.1 | - | 3 | 422 | |||
BT007348.1 | - | 3 | 289 | |||
DB272382.1 | - | 2 | 306 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
